accessibility menu, dialog, popup

UserName

CAS RN 221682-41-3

CH3OOHO(E/Z)

CAS RN 221682-41-3

IUPAC Name: 2-[3-oxo-2-(pent-2-en-1-yl)cyclopentyl]acetic acid
Synonyms: [3-oxo-2-(pent-2-en-1-yl)cyclopentyl]acetic acid
Molecular Weight (g/mol): 210.27
Molecular Formula: C12H18O3
InChi Key: ZNJFBWYDHIGLCU-UHFFFAOYNA-N
SMILES: CCC=CCC1C(CC(O)=O)CCC1=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results