missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Janus Green B, pure, high purity biological stain

CAS: 2869-83-2 | C30H31ClN6 | 511.07 g/mol

$139.79 - $139.79

Chemical Identifiers

CAS 2869-83-2
Molecular Formula C30H31ClN6
Molecular Weight (g/mol) 511.07
MDL Number MFCD00011758
InChI Key XXACTDWGHQXLGW-UHFFFAOYSA-M
Synonym C.I. 11050
PubChem CID 76123
ChEBI CHEBI:21184
IUPAC Name 8-[[4-(dimethylamino)phenyl]diazenyl]-N,N-diethyl-10-phenylphenazin-10-ium-2-amine;chloride
SMILES [Cl-].CCN(CC)C1=CC=C2N=C3C=CC(=CC3=[N+](C3=CC=CC=C3)C2=C1)N=NC1=CC=C(C=C1)N(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC191680050
SDS View Documents
Thermo Scientific Chemicals
191680050
5 g Glass bottle
Each for $139.79
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2869-83-2
511.07
XXACTDWGHQXLGW-UHFFFAOYSA-M
76123
8-[[4-(dimethylamino)phenyl]diazenyl]-N,N-diethyl-10-phenylphenazin-10-ium-2-amine;chloride
C30H31ClN6
MFCD00011758
C.I. 11050
CHEBI:21184
[Cl-].CCN(CC)C1=CC=C2N=C3C=CC(=CC3=[N+](C3=CC=CC=C3)C2=C1)N=NC1=CC=C(C=C1)N(C)C

Specifications

2869-83-2
C30H31ClN6
15, 5303
Solubility in water: soluble.
Authentic
8-[[4-(dimethylamino)phenyl]diazenyl]-N,N-diethyl-10-phenylphenazin-10-ium-2-amine;chloride
76123
511.07
Glass bottle
5 g
Janus Green B, Pure
50% min.
MFCD00011758
C.I. 11050
XXACTDWGHQXLGW-UHFFFAOYSA-M
[Cl-].CCN(CC)C1=CC=C2N=C3C=CC(=CC3=[N+](C3=CC=CC=C3)C2=C1)N=NC1=CC=C(C=C1)N(C)C
511.07
CHEBI:21184
Pure
Black or Brown/Green
Powder
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only