missing translation for 'onlineSavingsMsg'
Learn More

Synthware™ Jacketed Round Bottom Flask

Jacketed body to allow circulation of heating or cooling liquid

$1221.43 - $1338.10

Specifications

Color Clear
Material Borosilicate Glass (3.3)
For Use With (Application) Organic Synthesis
View More Specs

Products 4
Catalog Number Mfr. No. Capacity (Metric) Standard Taper Joint Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Standard Taper Joint Price Quantity  
31502162
View Documents
Synthware™
F20241L
1 L Center: 24/40; Side: 24/40
Each for $1,331.27
Only null left
 
31502164
View Documents
Synthware™
F20841L
1 L Center: 29/42; Side: 24/40
Each for $1,338.10
Only null left
 
31502163
View Documents
Synthware™
F204500
500 mL Center: 24/40; Side: 24/40
Each for $1,221.43
Only null left
 
31502165
View Documents
Synthware™
F208450
500 mL Center: 29/42; Side: 24/40
Each for $1,228.23
Only null left
 
Description

Description

  • Round bottom flask with standard taper outer joints on the center and vertical side necks
  • Heavy wall design
  • Entire flask body is jacketed and has two hose connections for circulation of heating or cooling liquid.
Specifications

Specifications

Clear
Borosilicate Glass (3.3)
Organic Synthesis
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'