Learn More
Ivermectin
CAS: 70288-86-7 | C48H74O14 | 875.106 g/mol
$61.96 - $158.36
Chemical Identifiers
| CAS | 70288-86-7 |
|---|---|
| Molecular Formula | C48H74O14 |
| Molecular Weight (g/mol) | 875.106 |
| MDL Number | MFCD00869511 |
| InChI Key | AZSNMRSAGSSBNP-RDKIRRGNSA-N |
| Synonym | ivermectin |
| PubChem CID | 122172888 |
| SMILES | CCC(C)C1C(CCC2(O1)CC3CC(O2)CC=C(C(C(C=CC=C4COC5C4(C(C=C(C5O)C)C(=O)O3)O)C)OC6CC(C(C(O6)C)OC7CC(C(C(O7)C)O)OC)OC)C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ62777MD
|
Thermo Scientific Chemicals
J62777MD |
250 mg |
Each for $61.96
|
|
|||||
|
AAJ6277703
|
Thermo Scientific Chemicals
J6277703 |
1 g |
Each for $158.36
|
|
|||||
Description
It is used as a positive allosteric modulator of the α7 neuronal nicotinic acetylcholine receptor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a positive allosteric modulator of the α7 neuronal nicotinic acetylcholine receptor.
Solubility
Soluble in methyl ethyl ketone (50 mg/ml), and DMSO (50 mM).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C.
Chemical Identifiers
| 70288-86-7 | |
| 875.106 | |
| AZSNMRSAGSSBNP-RDKIRRGNSA-N | |
| 122172888 |
| C48H74O14 | |
| MFCD00869511 | |
| ivermectin | |
| CCC(C)C1C(CCC2(O1)CC3CC(O2)CC=C(C(C(C=CC=C4COC5C4(C(C=C(C5O)C)C(=O)O3)O)C)OC6CC(C(C(O6)C)OC7CC(C(C(O7)C)O)OC)OC)C)C |
Specifications
| 70288-86-7 | |
| C48H74O14 | |
| 250 mg | |
| 14,5248 | |
| Soluble in methyl ethyl ketone (50mg/ml),and DMSO (50 mM). | |
| CCC(C)C1C(CCC2(O1)CC3CC(O2)CC=C(C(C(C=CC=C4COC5C4(C(C=C(C5O)C)C(=O)O3)O)C)OC6CC(C(C(O6)C)OC7CC(C(C(O7)C)O)OC)OC)C)C | |
| 122172888 | |
| Powder |
| White | |
| MFCD00869511 | |
| UN2811 | |
| ivermectin | |
| AZSNMRSAGSSBNP-RDKIRRGNSA-N | |
| 875.106 | |
| 875.11 | |
| Ivermectin |
Safety and Handling
GHS H Statement
H300-H360-H319
Fatal if swallowed.
May damage fertility or the unborn child.
Causes serious eye irritation.
P201-P202-P264b-P270-P280i-P281-P301+P310-P305+P351+P338-P308+P313-P330-P501c
H300-H319-H360
DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 274-536-0
RTECSNumber : IH7891500
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only