missing translation for 'onlineSavingsMsg'
Learn More

Ivermectin

CAS: 70288-86-7 | C48H74O14 | 875.106 g/mol

Supplier:  Thermo Scientific Chemicals J6277703

 View more versions of this product

Catalog No. AAJ6277703


Only null left
Explore available promotions

Description

Description

It is used as a positive allosteric modulator of the α7 neuronal nicotinic acetylcholine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a positive allosteric modulator of the α7 neuronal nicotinic acetylcholine receptor.

Solubility
Soluble in methyl ethyl ketone (50 mg/ml), and DMSO (50 mM).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C.
Specifications

Specifications

Ivermectin
White
MFCD00869511
UN2811
ivermectin
AZSNMRSAGSSBNP-RDKIRRGNSA-N
875.106
875.11
70288-86-7
C48H74O14
1 g
14,5248
Soluble in methyl ethyl ketone (50mg/ml),and DMSO (50 mM).
CCC(C)C1C(CCC2(O1)CC3CC(O2)CC=C(C(C(C=CC=C4COC5C4(C(C=C(C5O)C)C(=O)O3)O)C)OC6CC(C(C(O6)C)OC7CC(C(C(O7)C)O)OC)OC)C)C
122172888
Powder

Chemical Identifiers

70288-86-7
875.106
AZSNMRSAGSSBNP-RDKIRRGNSA-N
122172888
C48H74O14
MFCD00869511
ivermectin
CCC(C)C1C(CCC2(O1)CC3CC(O2)CC=C(C(C(C=CC=C4COC5C4(C(C=C(C5O)C)C(=O)O3)O)C)OC6CC(C(C(O6)C)OC7CC(C(C(O7)C)O)OC)OC)C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H360-H319
Fatal if swallowed.
May damage fertility or the unborn child.
Causes serious eye irritation.

P201-P202-P264b-P270-P280i-P281-P301+P310-P305+P351+P338-P308+P313-P330-P501c

H300-H319-H360

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 274-536-0

RTECSNumber : IH7891500

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only