missing translation for 'onlineSavingsMsg'
Learn More

Ivermectin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019437
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019437 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019437 Supplier Merck Emd Millipore Supplier No. PHR13801G
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Ivermectin is a derivative of avermectin B1 that is used against both endo- and ectoparasites in veterinary applications. It has also been indicated as a potential drug for the management of Onchocerca volvulus, identified as the cause of ′river blindness′ in humans. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 70288-86-7
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Physical Form Neat
MDL Number MFCD00869511
Synonym Ivermectin; 22,23-Dihydroavermectin B1; MK-933
Type Certified Reference Material
Quantity 1 g
Grade Certified Reference Material