missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ivabradine hydrochloride

Chemical Identifiers

CAS 148849-67-6
Chemical Name or Material Ivabradine hydrochloride
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC466280010
View Documents
Thermo Scientific™
466280010
1 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC466282500
View Documents
Thermo Scientific™
466282500
250 mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

148849-67-6
Ivabradine hydrochloride

Specifications

148849-67-6
Solubility aq. soln.: .. Other solubilities: soluble in dmso and methanol
Ivabradine hydrochloride
1 g
140.0°C
Videos
Safety and Handling

Safety and Handling

GHS H Statement Very toxic to aquatic life.

GHS P Statement Avoid release to the environment.

Collect spillage.

Dispose of contents/container to an approved waste disposal plant.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only