missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ivabradine hydrochloride

Supplier:  Thermo Scientific™ 466282500

 View more versions of this product

Catalog No. AC466282500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Ivabradine hydrochloride
250 mg
140.0°C
148849-67-6
Solubility aq. soln.: .. Other solubilities: soluble in dmso and methanol

Chemical Identifiers

148849-67-6
Ivabradine hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement Very toxic to aquatic life.

GHS P Statement Avoid release to the environment.

Collect spillage.

Dispose of contents/container to an approved waste disposal plant.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only