Learn More
Texwipe™ Non-Sterile Isopropyl Alcohol
Supplier:  Texwipe™ TX161
 View more versions of this product
Catalog No. 19-130-712
Only null left
Description
Description
Specifications
Specifications
Isopropyl Alcohol
100% Semiconductor-grade IPA
Ideal for surface cleaning, use on gloved hands, wipe down for pass through to controlled environments, use with cleanroom wipers, replaces solvent blending operations in facilities
0.47 L
Liquid
0.2 μm
16 oz.
Product Suggestions
Product Suggestions