missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ITSN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553702
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553702 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553702 Supplier Invitrogen™ Supplier No. PA553702
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: ITVLEQQDMW WFGEVQGQKG WFPKSYVKLI SGPIRKSTSM DSGSSESPAS LKRVASPAAK PVVSGEEFIA MYTYESSEQG DLTFQQGDVI LVTKKDGDWW Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 87%.

The protein encoded by this gene is a cytoplasmic membrane-associated protein that indirectly coordinates endocytic membrane traffic with the actin assembly machinery. In addition, the encoded protein may regulate the formation of clathrin-coated vesicles and could be involved in synaptic vesicle recycling. This protein has been shown to interact with dynamin, CDC42, SNAP23, SNAP25, SPIN90, EPS15, EPN1, EPN2, and STN2. Multiple transcript variants encoding different isoforms have been found for this gene, but the full-length nature of only two of them have been characterized so far.

Specifications

Antigen ITSN1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ITSN1
Gene Accession No. Q15811
Gene Alias AA517634; AA545208; AI316805; AI839402; AI848451; EH and SH3 domains protein 1; EH domain and SH3 domain regulator of endocytosis 1; Eh domain, SH3 domain regulator of endocytosis 1; Ehsh1; Ese1; human intersectin-SH3 domain-containing protein SH3P17; intersectin (SH3 domain protein 1A); intersectin 1; intersectin 1 (SH3 domain protein 1A); intersectin 1 (SH3 domain protein); intersectin 1 short form variant 3; intersectin 1 short form variant, 11; intersectin 1; intersectin-1; intersectin short form 2 variant 2; intersectin short variant 12; intersectin-1; intersectin-L; Itsn; ITSN1; MGC134948; MGC134949; SH3 domain-containing protein 1A; SH3D1A; SH3P17; Src homology 3 domain-containing protein
Gene Symbols ITSN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ITSN1. Recombinant protein control fragment (Product #RP-91801).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6453
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less