missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Itraconazole | |
| 169°C | |
| C35H38Cl2N8O4 | |
| 5 g | |
| VHVPQPYKVGDNFY-ZPGVKDDISA-N | |
| 1-(butan-2-yl)-4-{4-[4-(4-{[(2R,4S)-2-(2,4-dichlorophenyl)-2-[(1H-1,2,4-triazol-1-yl)methyl]-1,3-dioxolan-4-yl]methoxy}phenyl)piperazin-1-yl]phenyl}-4,5-dihydro-1H-1,2,4-triazol-5-one | |
| 45039617 | |
| ≥98.0% (HPLC,T) |
| 84625-61-6 | |
| White-Yellow | |
| MFCD00870168,MFCD00941396 | |
| itraconazole-d5, itraconazole, oriconazole-d5, triasporin-d5, sporamelt-d5, itrizole-d5, sporanox-d5, +/--4-4-4-4-2r,4s-2,4-dichlorophenyl-2-1h-1,2,4-triazol-1-ylmethyl-1,3-dioxolan-4-yl methoxy phenyl-1-piperazinyl phenyl-2,4-dihydro-2-1-methylpropyl-d5-3h-1,2,4-triazol-3-one, 2h5-4-4-4-4-2-1h-1,2,4-triazol-1-yl methyl-2-2,4-dichlorophenyl-1,3-dioxolan-4-yl methoxy phenyl piperazin-1-yl phenyl-1-sec-butyl-1h-1,2,4-triazol-5 4h-one, 2-3,3,4,4,4-2 h? butan-2-yl-4-4-4-4-2r,4s-2-2,4-dichlorophenyl-2-1,2,4-triazol-1-ylmethyl-1,3-dioxolan-4-yl methoxy phenyl piperazin-1-yl phenyl-1,2,4-triazol-3-one | |
| CCC(C)N1N=CN(C1=O)C1=CC=C(C=C1)N1CCN(CC1)C1=CC=C(OC[C@H]2CO[C@@](CN3C=NC=N3)(O2)C2=CC=C(Cl)C=C2Cl)C=C1 | |
| 705.64 | |
| 705.64 | |
| Crystalline Powder |
Chemical Identifiers
| 84625-61-6 | |
| 705.64 | |
| VHVPQPYKVGDNFY-ZPGVKDDISA-N | |
| 45039617 | |
| CCC(C)N1N=CN(C1=O)C1=CC=C(C=C1)N1CCN(CC1)C1=CC=C(OC[C@H]2CO[C@@](CN3C=NC=N3)(O2)C2=CC=C(Cl)C=C2Cl)C=C1 |
| C35H38Cl2N8O4 | |
| MFCD00870168,MFCD00941396 | |
| itraconazole-d5, itraconazole, oriconazole-d5, triasporin-d5, sporamelt-d5, itrizole-d5, sporanox-d5, +/--4-4-4-4-2r,4s-2,4-dichlorophenyl-2-1h-1,2,4-triazol-1-ylmethyl-1,3-dioxolan-4-yl methoxy phenyl-1-piperazinyl phenyl-2,4-dihydro-2-1-methylpropyl-d5-3h-1,2,4-triazol-3-one, 2h5-4-4-4-4-2-1h-1,2,4-triazol-1-yl methyl-2-2,4-dichlorophenyl-1,3-dioxolan-4-yl methoxy phenyl piperazin-1-yl phenyl-1-sec-butyl-1h-1,2,4-triazol-5 4h-one, 2-3,3,4,4,4-2 h? butan-2-yl-4-4-4-4-2r,4s-2-2,4-dichlorophenyl-2-1,2,4-triazol-1-ylmethyl-1,3-dioxolan-4-yl methoxy phenyl piperazin-1-yl phenyl-1,2,4-triazol-3-one | |
| 1-(butan-2-yl)-4-{4-[4-(4-{[(2R,4S)-2-(2,4-dichlorophenyl)-2-[(1H-1,2,4-triazol-1-yl)methyl]-1,3-dioxolan-4-yl]methoxy}phenyl)piperazin-1-yl]phenyl}-4,5-dihydro-1H-1,2,4-triazol-5-one |
Safety and Handling
RTECSNumber : XZ5481000
TSCA : No
Recommended Storage : Refrigerator