missing translation for 'onlineSavingsMsg'
Learn More

Itraconazole 98.0+%, TCI America™

Catalog No. p-7131567
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07325G 5 g
I073225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07325G Supplier TCI America Supplier No. I07325G

Chemical Identifiers

CAS 84625-61-6
Molecular Formula C35H38Cl2N8O4
Molecular Weight (g/mol) 705.64
MDL Number MFCD00870168,MFCD00941396
InChI Key VHVPQPYKVGDNFY-ZPGVKDDISA-N
Synonym itraconazole-d5, itraconazole, oriconazole-d5, triasporin-d5, sporamelt-d5, itrizole-d5, sporanox-d5, +/--4-4-4-4-2r,4s-2,4-dichlorophenyl-2-1h-1,2,4-triazol-1-ylmethyl-1,3-dioxolan-4-yl methoxy phenyl-1-piperazinyl phenyl-2,4-dihydro-2-1-methylpropyl-d5-3h-1,2,4-triazol-3-one, 2h5-4-4-4-4-2-1h-1,2,4-triazol-1-yl methyl-2-2,4-dichlorophenyl-1,3-dioxolan-4-yl methoxy phenyl piperazin-1-yl phenyl-1-sec-butyl-1h-1,2,4-triazol-5 4h-one, 2-3,3,4,4,4-2 h? butan-2-yl-4-4-4-4-2r,4s-2-2,4-dichlorophenyl-2-1,2,4-triazol-1-ylmethyl-1,3-dioxolan-4-yl methoxy phenyl piperazin-1-yl phenyl-1,2,4-triazol-3-one
PubChem CID 45039617
IUPAC Name 1-(butan-2-yl)-4-{4-[4-(4-{[(2R,4S)-2-(2,4-dichlorophenyl)-2-[(1H-1,2,4-triazol-1-yl)methyl]-1,3-dioxolan-4-yl]methoxy}phenyl)piperazin-1-yl]phenyl}-4,5-dihydro-1H-1,2,4-triazol-5-one
SMILES CCC(C)N1N=CN(C1=O)C1=CC=C(C=C1)N1CCN(CC1)C1=CC=C(OC[C@H]2CO[C@@](CN3C=NC=N3)(O2)C2=CC=C(Cl)C=C2Cl)C=C1

Specifications

Melting Point 169°C
Color White-Yellow
Quantity 5 g
Formula Weight 705.64
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Itraconazole
RTECSNumber XZ5481000
TSCA No
Recommended Storage Refrigerator