missing translation for 'onlineSavingsMsg'
Learn More

Itraconazole Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015670
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015670 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015670 Supplier Merck Emd Millipore Supplier No. PHR1834200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 84625-61-6
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C35H38 Cl2N8O4
MDL Number MFCD00870168
Synonym Itraconazole; Oriconazole; R51211; Sporanox
Type Certified Reference Material
Quantity 200 mg
Formula Weight 705.63
Grade Certified Reference Material