missing translation for 'onlineSavingsMsg'
Learn More

Itopride Hydrochloride 98.0+%, TCI America™

Catalog No. I09481G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09481G 1 g
I09485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09481G Supplier TCI America Supplier No. I09481G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 122892-31-3
Molecular Formula C20H27ClN2O4
Molecular Weight (g/mol) 394.896
MDL Number MFCD00881710
InChI Key ZTOUXLLIPWWHSR-UHFFFAOYSA-N
Synonym N-[4-[2-(Dimethylamino)ethoxy]benzyl]-3,4-dimethoxybenzamide Hydrochloride
PubChem CID 129791
IUPAC Name N-[[4-[2-(dimethylamino)ethoxy]phenyl]methyl]-3,4-dimethoxybenzamide;hydrochloride
SMILES CN(C)CCOC1=CC=C(C=C1)CNC(=O)C2=CC(=C(C=C2)OC)OC.Cl

Specifications

Melting Point 194°C
Color White
Quantity 1 g
Formula Weight 394.90
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Itopride Hydrochloride
RTECSNumber CV4552000
TSCA No
Recommended Storage Refrigerator