missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITGB1BP2 Monoclonal Antibody (K52028_12C5), Invitrogen™
Mouse Monoclonal Antibody
$1,601.06 - $2,821.92
Specifications
| Antigen | ITGB1BP2 |
|---|---|
| Clone | K52028_12C5 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: EAPQPEGPAT SSSLQEQKPL NVIPKSAETL RRERPKSELP LKLLPLNISQ ALEMALEQKE LDQEPGAGLD SLIRTGSSCQ NPGCDAVYQG PESDATPCTY HPGAPRFHEG MKSWSCCGIQ TLDFGAFLAQ PGCRVGRHDW GKQLPASCRH DWHQTDSLVV VTVYGQIPLP AFNWVKASQT ELHVHIVFDG NRVFQAQMKL WGVINVEQSS VFLMPSRVEI SLVKADPGSW AQLEHPDALA KKARAGVVLE MDEEESDDSD DDLSWTEEEE
ITGB1BP2 Monoclonal antibody specifically detects ITGB1BP2 in Human, Rat samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| ITGB1BP2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Chordc3; integrin beta 1 binding protein (melusin) 2; integrin beta 1 binding protein 2; integrin beta-1-binding protein 2; integrin subunit beta 1 binding protein 2; ITGB1BP; Itgb1bp2; Melusin; MGC119214; MSTP015; RGD1565015 | |
| Recombinant human ITGB1BP2 (immunogen range is 71-340). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| ITGB1BP2 |
| K52028_12C5 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| Q9UKP3 | |
| 26548, 317258 | |
| IgG1 | |
| Protein A | |
| Antibody |