missing translation for 'onlineSavingsMsg'
Learn More

Itaconic anhydride, 97%

CAS: 2170-03-8 | C5H4O3 | 112.08 g/mol

Supplier:  Thermo Scientific Chemicals B2521622

 View more versions of this product

Catalog No. AAB2521622


Only null left
Explore available promotions

Description

Description

Itaconic anhydride is used in immobilization of β-glucosidase. It undergoes estrification to obtain suitable monomers for emulsion polymerization itaconates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Itaconic anhydride is used in immobilization of β-glucosidase. It undergoes estrification to obtain suitable monomers for emulsion polymerization itaconates.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents and moisture.
Specifications

Specifications

Itaconic anhydride
2170-03-8
100 g
MFCD00005530
itaconic anhydride, itaconic acid anhydride, 2-methylenesuccinic anhydride, methylenesuccinic anhydride, 2,5-furandione, dihydro-3-methylene, itaconicanhydride, succinic anhydride, methylene, dihydro-3-methylene-2,5-furandione, 3-methylenedihydro-2,5-furandione, unii-y455ks1u7q
OFNISBHGPNMTMS-UHFFFAOYSA-N
3-methylideneoxolane-2,5-dione
75110
97%
68°C to 72°C
114°C to 115°C (12 mmHg)
C5H4O3
2212
Hydrolyzes in water.
C=C1CC(=O)OC1=O
112.08
112.09

Chemical Identifiers

2170-03-8
112.08
OFNISBHGPNMTMS-UHFFFAOYSA-N
75110
C=C1CC(=O)OC1=O
C5H4O3
MFCD00005530
itaconic anhydride, itaconic acid anhydride, 2-methylenesuccinic anhydride, methylenesuccinic anhydride, 2,5-furandione, dihydro-3-methylene, itaconicanhydride, succinic anhydride, methylene, dihydro-3-methylene-2,5-furandione, 3-methylenedihydro-2,5-furandione, unii-y455ks1u7q
3-methylideneoxolane-2,5-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P280-P305+P351+P338-P310

H318

EINECSNumber : 218-518-2

TSCA : Yes

Recommended Storage : Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only