missing translation for 'onlineSavingsMsg'
Learn More

Itaconic anhydride, 97%

Catalog No. p-7028088
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2521614 25 g
AAB2521622 100 g
AAB2521636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2521614 Supplier Thermo Scientific Chemicals Supplier No. B2521614
Only null left

CAS: 2170-03-8 | C5H4O3 | 112.08 g/mol

Itaconic anhydride is used in immobilization of β-glucosidase. It undergoes estrification to obtain suitable monomers for emulsion polymerization itaconates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Itaconic anhydride is used in immobilization of β-glucosidase. It undergoes estrification to obtain suitable monomers for emulsion polymerization itaconates.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents and moisture.

Chemical Identifiers

CAS 2170-03-8
Molecular Formula C5H4O3
Molecular Weight (g/mol) 112.08
MDL Number MFCD00005530
InChI Key OFNISBHGPNMTMS-UHFFFAOYSA-N
Synonym itaconic anhydride, itaconic acid anhydride, 2-methylenesuccinic anhydride, methylenesuccinic anhydride, 2,5-furandione, dihydro-3-methylene, itaconicanhydride, succinic anhydride, methylene, dihydro-3-methylene-2,5-furandione, 3-methylenedihydro-2,5-furandione, unii-y455ks1u7q
PubChem CID 75110
IUPAC Name 3-methylideneoxolane-2,5-dione
SMILES C=C1CC(=O)OC1=O

Specifications

Melting Point 68°C to 72°C
Boiling Point 114°C to 115°C (12 mmHg)
Quantity 25 g
Beilstein 2212
Solubility Information Hydrolyzes in water.
Formula Weight 112.09
Percent Purity 97%
Chemical Name or Material Itaconic anhydride
Hazard Category H318
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P280-P305+P351+P338-P310
EINECSNumber 218-518-2
TSCA Yes
Recommended Storage Store under Nitrogen

RUO – Research Use Only