missing translation for 'onlineSavingsMsg'
Learn More

Isoxazolidine Hydrochloride 97.0+%, TCI America™

Catalog No. I09661G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09661G 1 g
I09665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09661G Supplier TCI America Supplier No. I09661G
Only null left

Chemical Identifiers

CAS 39657-45-9
Molecular Formula C3H8ClNO
Molecular Weight (g/mol) 109.553
MDL Number MFCD11100465
InChI Key HPIVVOQIZYNVJE-UHFFFAOYSA-N
PubChem CID 12304500
IUPAC Name 1,2-oxazolidine;hydrochloride
SMILES C1CNOC1.Cl

Specifications

Melting Point 127°C
Color White
Quantity 1 g
Formula Weight 109.55
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Isoxazolidine Hydrochloride