accessibility menu, dialog, popup

UserName

NHOHCl

CAS RN 39657-45-9

IUPAC Name: 1,2-oxazolidine hydrochloride
Synonyms: isoxazolidine hydrochloride
Molecular Weight (g/mol): 109.55
Molecular Formula: C3H8ClNO
InChi Key: HPIVVOQIZYNVJE-UHFFFAOYSA-N
SMILES: Cl.C1CNOC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results