missing translation for 'onlineSavingsMsg'
Learn More

Isoxazole, 99%

Catalog No. aal0160706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0160706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0160706 Supplier Thermo Scientific Chemicals Supplier No. L0160706
Only null left

CAS: 288-14-2 | C3H3NO | 69.063 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 288-14-2
Molecular Formula C3H3NO
Molecular Weight (g/mol) 69.063
MDL Number MFCD00003149
InChI Key CTAPFRYPJLPFDF-UHFFFAOYSA-N
Synonym isoxazole, 1-oxa-2-azacyclopentadiene, isooxazole, unii-00srw0m6pw, 00srw0m6pw, isoxazoles, pubchem8623, acmc-209h4x, ksc204o6n
PubChem CID 9254
ChEBI CHEBI:35595
IUPAC Name 1,2-oxazole
SMILES C1=CON=C1

Specifications

Density 1.078
Boiling Point 93°C to 95°C
Flash Point 8°C (46°F)
Refractive Index 1.427
Quantity 5 g
UN Number UN1993
Beilstein 103773
Formula Weight 69.06
Percent Purity 99%
Chemical Name or Material Isoxazole
Hazard Category H225-H314-H335
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 206-018-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only