accessibility menu, dialog, popup

UserName

CAS RN 288-14-2

IUPAC Name: 1,2-oxazole
Synonyms: isoxazole 1-oxa-2-azacyclopentadiene
Molecular Weight (g/mol): 69.06
Molecular Formula: C3H3NO
InChi Key: CTAPFRYPJLPFDF-UHFFFAOYSA-N
SMILES: O1C=CC=N1
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)