Learn More
Isoxazole-4-boronic acid pinacol ester, 97%
CAS: 928664-98-6 | C9H14BNO3 | 195.025 g/mol
Supplier: Thermo Scientific Chemicals H6688703
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Isoxazole-4-boronic acid pinacol ester | |
| 110°C to 115°C | |
| MFCD06657891 | |
| 4-isoxazoleboronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl isoxazole, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-isoxazole, isoxazol-4-ylboronic acid pinacol ester, isoxazole-4-boronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2-oxazole, 4-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2-oxazole, isoxazole, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-isooxazolylboronic acid pinacol ester, 4-isoxazoleboronicacidpinacolester | |
| B1(OC(C(O1)(C)C)(C)C)C2=CON=C2 | |
| 195.025 | |
| 195.02 |
| 928664-98-6 | |
| C9H14BNO3 | |
| 1 g | |
| LXCICYRNWIGDQA-UHFFFAOYSA-N | |
| 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2-oxazole | |
| 16414180 | |
| 97% |
Chemical Identifiers
| 928664-98-6 | |
| 195.025 | |
| LXCICYRNWIGDQA-UHFFFAOYSA-N | |
| 16414180 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CON=C2 |
| C9H14BNO3 | |
| MFCD06657891 | |
| 4-isoxazoleboronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl isoxazole, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-isoxazole, isoxazol-4-ylboronic acid pinacol ester, isoxazole-4-boronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2-oxazole, 4-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2-oxazole, isoxazole, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-isooxazolylboronic acid pinacol ester, 4-isoxazoleboronicacidpinacolester | |
| 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2-oxazole |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only