missing translation for 'onlineSavingsMsg'
Learn More

Isoxadifen-ethyl 98.0+%, TCI America™

Catalog No. I09565G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09565G 5 g
I095625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09565G Supplier TCI America Supplier No. I09565G

Chemical Identifiers

CAS 163520-33-0
Molecular Formula C18H17NO3
Molecular Weight (g/mol) 295.34
MDL Number MFCD03792846
InChI Key MWKVXOJATACCCH-UHFFFAOYSA-N
Synonym isoxadifen-ethyl, ethyl 5,5-diphenyl-4,5-dihydroisoxazole-3-carboxylate, isoxadifen-ethyl iso:bsi, unii-71lf32w32e, 3-isoxazolecarboxylic acid, 4,5-dihydro-5,5-diphenyl-, ethyl ester, 4,5-dihydro-5,5-diphenyl-1,2-oxazole-3-carboxylic acid, ethyl ester, ethyl 5,5-diphenyl-2-isoxazoline-3-carboxylate, isoxadifen etil, isoxadifen ethyl, dsstox_cid_20360
PubChem CID 6451155
IUPAC Name ethyl 5,5-diphenyl-4,5-dihydro-1,2-oxazole-3-carboxylate
SMILES CCOC(=O)C1=NOC(C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 87°C
Color White
Quantity 5 g
UN Number 3077
Formula Weight 295.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Isoxadifen-ethyl
RTECSNumber NY2390000
TSCA Yes