missing translation for 'onlineSavingsMsg'
Learn More

Isovalerophenone 99.0+%, TCI America™

Catalog No. I02965ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I02965ML 5 mL
I029625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I02965ML Supplier TCI America Supplier No. I02965ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 582-62-7
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
MDL Number MFCD00026486
InChI Key HEOVGVNITGAUKL-UHFFFAOYSA-N
Synonym Isobutyl Phenyl Ketone
PubChem CID 68493
IUPAC Name 3-methyl-1-phenylbutan-1-one
SMILES CC(C)CC(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 236°C
Quantity 5 mL
Formula Weight 162.23
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Isovalerophenone