CAS RN 582-62-7
IUPAC Name:
3-methyl-1-phenylbutan-1-one
Synonyms:
1-butanone, 3-methyl-1-phenyl-
Molecular Weight (g/mol):
162.23
Molecular Formula:
C11H14O
InChi Key:
HEOVGVNITGAUKL-UHFFFAOYSA-N
SMILES:
CC(C)CC(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
Isovalerophenone 99.0+%, TCI America™
CAS: 582-62-7 Molecular Formula: C11H14O Molecular Weight (g/mol): 162.23 MDL Number: MFCD00026486 InChI Key: HEOVGVNITGAUKL-UHFFFAOYSA-N Synonym: Isobutyl Phenyl Ketone PubChem CID: 68493 IUPAC Name: 3-methyl-1-phenylbutan-1-one SMILES: CC(C)CC(=O)C1=CC=CC=C1