missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isovaleronitrile 98.0+%, TCI America™
$232.95 - $1524.74
Chemical Identifiers
| CAS | 625-28-5 |
|---|---|
| Molecular Formula | C5H9N |
| Molecular Weight (g/mol) | 83.134 |
| MDL Number | MFCD00001944 |
| InChI Key | QHDRKFYEGYYIIK-UHFFFAOYSA-N |
| Synonym | isovaleronitrile, butanenitrile, 3-methyl, isoamylnitrile, isobutyl cyanide, isopentane nitrile, 3-methylbutyronitrile, butyronitrile, 3-methyl, 2-methylbutane secondary mononitrile, unii-n2g72x091l, isopentanenitrile |
| PubChem CID | 12244 |
| IUPAC Name | 3-methylbutanenitrile |
| SMILES | CC(C)CC#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
I029225ML
|
TCI America
I029225ML |
25 mL |
Each for $232.95
|
|
|||||
|
I0292100ML
|
TCI America
I0292100ML |
100 mL |
Each for $508.25
|
|
|||||
|
I0292500ML
|
TCI America
I0292500ML |
500 mL |
Each for $1,524.74
|
|
|||||
Chemical Identifiers
| 625-28-5 | |
| 83.134 | |
| QHDRKFYEGYYIIK-UHFFFAOYSA-N | |
| 12244 | |
| CC(C)CC#N |
| C5H9N | |
| MFCD00001944 | |
| isovaleronitrile, butanenitrile, 3-methyl, isoamylnitrile, isobutyl cyanide, isopentane nitrile, 3-methylbutyronitrile, butyronitrile, 3-methyl, 2-methylbutane secondary mononitrile, unii-n2g72x091l, isopentanenitrile | |
| 3-methylbutanenitrile |
Specifications
| 625-28-5 | |
| 130°C | |
| MFCD00001944 | |
| 1992 | |
| QHDRKFYEGYYIIK-UHFFFAOYSA-N | |
| 3-methylbutanenitrile | |
| 12244 | |
| ≥98.0% (GC) | |
| Isovaleronitrile |
| Colorless | |
| C5H9N | |
| 25 mL | |
| isovaleronitrile, butanenitrile, 3-methyl, isoamylnitrile, isobutyl cyanide, isopentane nitrile, 3-methylbutyronitrile, butyronitrile, 3-methyl, 2-methylbutane secondary mononitrile, unii-n2g72x091l, isopentanenitrile | |
| CC(C)CC#N | |
| 83.134 | |
| 83.13 | |
| Liquid |
Safety and Handling
RTECSNumber : NY1750000
TSCA : Yes