accessibility menu, dialog, popup

UserName

CAS RN 625-28-5

IUPAC Name: 3-methylbutanenitrile
Synonyms: butanenitrile, 3-methyl-
Molecular Weight (g/mol): 83.13
Molecular Formula: C5H9N
InChi Key: QHDRKFYEGYYIIK-UHFFFAOYSA-N
SMILES: CC(C)CC#N
Boiling Point 
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)