missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isovaleronitrile 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 130°C |
| Quantity | 25 mL |
| UN Number | 1992 |
| Formula Weight | 83.13 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Isovaleronitrile |
Safety and Handling
| RTECSNumber | NY1750000 |
| TSCA | Yes |