Learn More
Isovaleronitrile, 98%
CAS: 625-28-5 | C5H9N | 83.13 g/mol
Supplier: Thermo Scientific Chemicals 427560050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Isovaleronitrile | |
| Colorless | |
| 128.0°C to 130.0°C (730.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| (CH3)2CHCH2CN | |
| 5 g | |
| isovaleronitrile, butanenitrile, 3-methyl, isoamylnitrile, isobutyl cyanide, isopentane nitrile, 3-methylbutyronitrile, butyronitrile, 3-methyl, 2-methylbutane secondary mononitrile, unii-n2g72x091l, isopentanenitrile | |
| CC(C)CC#N | |
| 83.13 | |
| 83.13 | |
| Liquid |
| 625-28-5 | |
| 0.7950g/mL | |
| 28°C | |
| 97.5% min. (GC) | |
| C5H9N | |
| MFCD00001944 | |
| 0.795 | |
| QHDRKFYEGYYIIK-UHFFFAOYSA-N | |
| 3-methylbutanenitrile | |
| 12244 | |
| 98% |
Chemical Identifiers
| 625-28-5 | |
| 83.13 | |
| QHDRKFYEGYYIIK-UHFFFAOYSA-N | |
| 12244 | |
| CC(C)CC#N |
| C5H9N | |
| MFCD00001944 | |
| isovaleronitrile, butanenitrile, 3-methyl, isoamylnitrile, isobutyl cyanide, isopentane nitrile, 3-methylbutyronitrile, butyronitrile, 3-methyl, 2-methylbutane secondary mononitrile, unii-n2g72x091l, isopentanenitrile | |
| 3-methylbutanenitrile |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF
GHS Signal Word: Danger
EINECSNumber : 210-884-1
Recommended Storage : 4°C
RUO – Research Use Only