missing translation for 'onlineSavingsMsg'
Learn More

Isovaleraldehyde, ≥97% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019860
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019860 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019860 Supplier Merck Emd Millipore Supplier No. 61848100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 590-86-3
Ignition Point 464°F
Vapor Pressure 30 mmHg (20°C)
Vapor Density 2.96 (vs. air)
Density 0.803 g/mL (at 25°C)
Flash Point 0.5°C - closed cup
Physical Form Neat
Linear Formula (CH3)2 CHCH2 CHO
MDL Number MFCD00007014
Boiling Point 90°C
Synonym 3-Methylbutanal; 3-Methylbutyraldehyde; NSC 404119
Type Analytical Standard
Quantity 100 mg
Formula Weight 86.13
Percent Purity ≥97% (GC)
Grade Analytical Standard
Show More Show Less