missing translation for 'onlineSavingsMsg'
Learn More

Isothiazole, 97%, Thermo Scientific Chemicals

Catalog No. AC458420010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458420010 1 g
AC458420050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC458420010 Supplier Thermo Scientific Chemicals Supplier No. 458420010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 288-16-4
Molecular Formula C3H3NS
Molecular Weight (g/mol) 85.13
InChI Key ZLTPDFXIESTBQG-UHFFFAOYSA-N
Synonym isothiazole, 2-azathiophene, 1,2-thiazol, isothiazole;1,2-thiazole, 4-fluoro-3'-methylbenzophenone
PubChem CID 67515
ChEBI CHEBI:35600
IUPAC Name 1,2-thiazole
SMILES C1=CSN=C1

Specifications

Flash Point 22°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 85.13
Percent Purity 97%
Chemical Name or Material Isothiazole
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Highly flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation.
Precautionary Statement GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wash face, hands and any exposed skin thoroughly after handling. If skin irritation occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only