missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isosorbide Solution, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™
Description
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Isosorbide (Dianhydro-D-glucitol) is a greener ingredient widely used as a monomer and building block for a variety of polymers and functional materials, for medical and pharmaceutical applications. It is also used as a fuel or fuel additive.
Specifications
Specifications
| Linear Formula | C6H10O4 |
| Synonym | Dianhydro-D-glucitol solution |
| Quantity | 1 g |
| Formula Weight | 146.14 |
| Grade | Certified Reference Material |