missing translation for 'onlineSavingsMsg'
Learn More

Isoquinoline-5-boronic acid, 97%

Catalog No. p-7035513
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH53230MD 250 mg
AAH5323003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH53230MD Supplier Thermo Scientific Chemicals Supplier No. H53230MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 371766-08-4 | C9H8BNO2 | 172.978 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 371766-08-4
Molecular Formula C9H8BNO2
Molecular Weight (g/mol) 172.978
MDL Number MFCD03839356
InChI Key XKEYHBLSCGBBGU-UHFFFAOYSA-N
Synonym isoquinoline-5-boronic acid, 5-isoquinolineboronic acid, isoquinolin-5-yl-5-boronic acid, boronic acid, 5-isoquinolinyl, isoquinolin-5-yl boronic acid, 5-isoquinolineboronicacid, isoquinoline-5-boric acid, 5-boronoisoquinoline, acmc-209ird, 5-isoquinolylboronic acid
PubChem CID 599474
IUPAC Name isoquinolin-5-ylboronic acid
SMILES B(C1=C2C=CN=CC2=CC=C1)(O)O

Specifications

Melting Point 204°C to 206°C
Quantity 250 mg
Formula Weight 172.98
Percent Purity 97%
Chemical Name or Material Isoquinoline-5-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only