CAS RN 371766-08-4
IUPAC Name:
(isoquinolin-5-yl)boronic acid
Synonyms:
isoquinolin-5-ylboronic acid
Molecular Weight (g/mol):
172.98
Molecular Formula:
C9H8BNO2
InChi Key:
XKEYHBLSCGBBGU-UHFFFAOYSA-N
SMILES:
OB(O)C1=C2C=CN=CC2=CC=C1
1
–
1
of
1
results
1
Promotions Available
Isoquinoline-5-boronic acid, 97%
Isoquinoline-5-boronic acid, 97%
CAS: 371766-08-4 Molecular Formula: C9H8BNO2 Molecular Weight (g/mol): 172.978 MDL Number: MFCD03839356 InChI Key: XKEYHBLSCGBBGU-UHFFFAOYSA-N Synonym: isoquinoline-5-boronic acid,5-isoquinolineboronic acid,isoquinolin-5-yl-5-boronic acid,boronic acid, 5-isoquinolinyl,isoquinolin-5-yl boronic acid,5-isoquinolineboronicacid,isoquinoline-5-boric acid,5-boronoisoquinoline,acmc-209ird,5-isoquinolylboronic acid PubChem CID: 599474 IUPAC Name: isoquinolin-5-ylboronic acid SMILES: B(C1=C2C=CN=CC2=CC=C1)(O)O