missing translation for 'onlineSavingsMsg'
Learn More

Isoquinoline-4-carboxaldehyde, 97%

Catalog No. AAH6178803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6178803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6178803 Supplier Thermo Scientific Chemicals Supplier No. H6178803
Only null left

CAS: 22960-16-3 | C10H7NO | 157.172 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22960-16-3
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.172
MDL Number MFCD00829440
InChI Key RNQQJLYJLDQGGL-UHFFFAOYSA-N
Synonym isoquinoline-4-carboxaldehyde, 4-isoquinolinecarboxaldehyde, 4-formylisoquinoline, pubchem15962, acmc-20aig4, 4-isoquinolinecarbaldehyde, ksc201k4f, 4-isoquinolinecarboxaldehyde 7ci,8ci,9ci
PubChem CID 10868870
IUPAC Name isoquinoline-4-carbaldehyde
SMILES C1=CC=C2C(=C1)C=NC=C2C=O

Specifications

Melting Point 101°C to 106°C
Quantity 1 g
Formula Weight 157.17
Percent Purity 97%
Chemical Name or Material Isoquinoline-4-carboxaldehyde
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H318
Toxic if swallowed.
Causes serious eye damage.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only