accessibility menu, dialog, popup

UserName

NO

CAS RN 22960-16-3

IUPAC Name: isoquinoline-4-carbaldehyde
Synonyms: isoquinoline-4-carbaldehyde
Molecular Weight (g/mol): 157.17
Molecular Formula: C10H7NO
InChi Key: RNQQJLYJLDQGGL-UHFFFAOYSA-N
SMILES: O=CC1=CN=CC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results