missing translation for 'onlineSavingsMsg'
Learn More

Isoquinoline-3-carboxylic Acid Monohydrate 98.0+%, TCI America™

Catalog No. I05091G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05091G 1 g
I05095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05091G Supplier TCI America Supplier No. I05091G
Only null left

Chemical Identifiers

CAS 203626-75-9
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.186
MDL Number MFCD00075137
InChI Key IXFLTVWYYQIVRB-UHFFFAOYSA-N
Synonym isoquinoline-3-carboxylic acid hydrate, 3-isoquinolinecarboxylic acid hydrate, isoquinoline-3-carboxylic acid monohydrate, isoquinoline-3-carboxylic acid hydrate 1:x, 3-isoquinolinecarboxylic acid, hydrate, 3-isoquinolinecarboxylic acid hydrate 1:1, pubchem10167, acmc-1clhb, 3-isoquinolinecarboxylicacid hydrate, isoquinoline 3-carboxylic acid hydrate
PubChem CID 2736704
IUPAC Name isoquinoline-3-carboxylic acid;hydrate
SMILES C1=CC=C2C=NC(=CC2=C1)C(=O)O.O

Specifications

Melting Point 167°C
Quantity 1 g
Formula Weight 173.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isoquinoline-3-carboxylic Acid Monohydrate