missing translation for 'onlineSavingsMsg'
Learn More

Isoquinoline-3-carboxylic acid hydrate, 99%

CAS: 207399-25-5 | C10H7NO2 | 173.17 g/mol

Supplier:  Thermo Scientific Chemicals A1988006

 View more versions of this product

Catalog No. AAA1988006

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Isoquinoline-3-carboxylic acid hydrate
166°C to 168°C
MFCD00075137
125548
KVMMIDQDXZOPAB-UHFFFAOYSA-N
isoquinoline-3-carboxylic acid
2736704
99%
207399-25-5
C10H7NO2
5 g
isoquinoline-3-carboxylic acid hydrate, 3-isoquinolinecarboxylic acid hydrate, isoquinoline-3-carboxylic acid monohydrate, isoquinoline-3-carboxylic acid hydrate 1:x, 3-isoquinolinecarboxylic acid, hydrate, 3-isoquinolinecarboxylic acid hydrate 1:1, pubchem10167, acmc-1clhb, 3-isoquinolinecarboxylicacid hydrate, isoquinoline 3-carboxylic acid hydrate
OC(=O)C1=CC2=CC=CC=C2C=N1
173.17
173.17 (Anhydrous)

Chemical Identifiers

207399-25-5
173.17
KVMMIDQDXZOPAB-UHFFFAOYSA-N
2736704
C10H7NO2
MFCD00075137
isoquinoline-3-carboxylic acid hydrate, 3-isoquinolinecarboxylic acid hydrate, isoquinoline-3-carboxylic acid monohydrate, isoquinoline-3-carboxylic acid hydrate 1:x, 3-isoquinolinecarboxylic acid, hydrate, 3-isoquinolinecarboxylic acid hydrate 1:1, pubchem10167, acmc-1clhb, 3-isoquinolinecarboxylicacid hydrate, isoquinoline 3-carboxylic acid hydrate
OC(=O)C1=CC2=CC=CC=C2C=N1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only