missing translation for 'onlineSavingsMsg'
Learn More

Isoquinoline-3-carboxylic acid hydrate, 99%

Catalog No. p-7024712
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1988003 1 g
AAA1988006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1988003 Supplier Thermo Scientific Chemicals Supplier No. A1988003
Only null left

CAS: 207399-25-5 | C10H7NO2 | 173.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 207399-25-5
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.17
MDL Number MFCD00075137
InChI Key KVMMIDQDXZOPAB-UHFFFAOYSA-N
Synonym isoquinoline-3-carboxylic acid hydrate, 3-isoquinolinecarboxylic acid hydrate, isoquinoline-3-carboxylic acid monohydrate, isoquinoline-3-carboxylic acid hydrate 1:x, 3-isoquinolinecarboxylic acid, hydrate, 3-isoquinolinecarboxylic acid hydrate 1:1, pubchem10167, acmc-1clhb, 3-isoquinolinecarboxylicacid hydrate, isoquinoline 3-carboxylic acid hydrate
PubChem CID 2736704
SMILES OC(=O)C1=CC2=CC=CC=C2C=N1

Specifications

Melting Point 166°C to 168°C
Quantity 1 g
Beilstein 125548
IUPAC Name isoquinoline-3-carboxylic acid;hydrate
Formula Weight 173.17 (Anhydrous)
Percent Purity 99%
Chemical Name or Material Isoquinoline-3-carboxylic acid hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only