missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoquinolin-3-amine, 97%, Thermo Scientific™
$95.96 - $95.96
Chemical Identifiers
| CAS | 25475-67-6 |
|---|---|
| Molecular Formula | C9H8N2 |
| Molecular Weight (g/mol) | 144.177 |
| InChI Key | VYCKDIRCVDCQAE-UHFFFAOYSA-N |
| Synonym | 3-aminoisoquinoline, 3-isoquinolinamine, isoquinolin-3-ylamine, isoquinoline, 3-amino, 3-isoquinolylamine, isoquinoline-3-amine, chembl61743, 2h-isoquinolin-3-imine, isoquinolin-3-imine, zlchem 309 |
| PubChem CID | 311869 |
| IUPAC Name | isoquinolin-3-amine |
| SMILES | C1=CC=C2C=NC(=CC2=C1)N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
BTB10019CB
|
Thermo Scientific Chemicals
BTB10019CB |
250 mg | Amber Glass Bottle |
Each for $95.96
|
|
||||
Chemical Identifiers
| 25475-67-6 | |
| 144.177 | |
| 3-aminoisoquinoline, 3-isoquinolinamine, isoquinolin-3-ylamine, isoquinoline, 3-amino, 3-isoquinolylamine, isoquinoline-3-amine, chembl61743, 2h-isoquinolin-3-imine, isoquinolin-3-imine, zlchem 309 | |
| isoquinolin-3-amine |
| C9H8N2 | |
| VYCKDIRCVDCQAE-UHFFFAOYSA-N | |
| 311869 | |
| C1=CC=C2C=NC(=CC2=C1)N |
Specifications
| 25475-67-6 | |
| Amber Glass Bottle | |
| 250 mg | |
| VYCKDIRCVDCQAE-UHFFFAOYSA-N | |
| isoquinolin-3-amine | |
| 311869 | |
| 97% |
| 97% | |
| C9H8N2 | |
| 3-aminoisoquinoline, 3-isoquinolinamine, isoquinolin-3-ylamine, isoquinoline, 3-amino, 3-isoquinolylamine, isoquinoline-3-amine, chembl61743, 2h-isoquinolin-3-imine, isoquinolin-3-imine, zlchem 309 | |
| C1=CC=C2C=NC(=CC2=C1)N | |
| 144.177 | |
| 144.18 | |
| Isoquinolin-3-amine |