CAS RN 25475-67-6
CAS RN 25475-67-6
IUPAC Name:
isoquinolin-3-amine
Synonyms:
isoquinolin-3-amine
Molecular Weight (g/mol):
144.18
Molecular Formula:
C9H8N2
InChi Key:
VYCKDIRCVDCQAE-UHFFFAOYSA-N
SMILES:
NC1=NC=C2C=CC=CC2=C1
1
–
2
of
2
results
1
Isoquinolin-3-amine, 97%, Thermo Scientific™
CAS: 25475-67-6 Molecular Formula: C9H8N2 Molecular Weight (g/mol): 144.177 InChI Key: VYCKDIRCVDCQAE-UHFFFAOYSA-N Synonym: 3-aminoisoquinoline,3-isoquinolinamine,isoquinolin-3-ylamine,isoquinoline, 3-amino,3-isoquinolylamine,isoquinoline-3-amine,chembl61743,2h-isoquinolin-3-imine,isoquinolin-3-imine,zlchem 309 PubChem CID: 311869 IUPAC Name: isoquinolin-3-amine SMILES: C1=CC=C2C=NC(=CC2=C1)N