missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Isopulegol, Analytical Standard, ≥99.0% (sum of enantiomers, GC), MilliporeSigma™ Supelco™
Description
Monoterpenoid chiral building block, hydroboration to a 1,4-diol
Specifications
Specifications
| CAS | 89-79-2 |
| Density | 0.912 g/mL (at 25°C) |
| Physical Form | Neat |
| Linear Formula | C10H18O |
| MDL Number | MFCD00134655 |
| Boiling Point | 212°C |
| Synonym | (1 R,2 S,5 R)-2-Isopropenyl-5-methylcyclohexanol; (1 R,3 R,4 S)-p-Menth-8-en-3-ol |
| Quantity | 5 mL |
| Formula Weight | 154.25 |
| Percent Purity | ≥99.0% (sum of enantiomers, GC) |
| Show More |