missing translation for 'onlineSavingsMsg'
Learn More

(+)-Isopulegol, Analytical Standard, ≥99.0% (sum of enantiomers, GC), MilliporeSigma™ Supelco™

Catalog No. 111018793
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018793 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018793 Supplier Merck Emd Millipore Supplier No. 597651ML
Only null left

Specifications

CAS 104870-56-6
Density 0.912 g/mL (at 25°C)
Physical Form Neat
Linear Formula C10H18O
MDL Number MFCD00213790
Boiling Point 91°C (12 mmHg)
Synonym (1 S,2 R,5 S)-2-Isopropenyl-5-methylcyclohexanol; (1 S,3 S,4 R)-p-Menth-8-en-3-ol
Quantity 1 mL
Formula Weight 154.25
Percent Purity ≥99.0% (sum of enantiomers, GC)
Grade Analytical Standard
Show More Show Less