missing translation for 'onlineSavingsMsg'
Learn More

Isopropylsulfonyl Chloride 95.0+%, TCI America™

Catalog No. p-7131311
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I02915G 5 g
I029125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I02915G Supplier TCI America Supplier No. I02915G
Only null left

Chemical Identifiers

CAS 10147-37-2
Molecular Formula C3H7ClO2S
Molecular Weight (g/mol) 142.597
MDL Number MFCD00007453
InChI Key DRINJBFRTLBHNF-UHFFFAOYSA-N
Synonym isopropylsulfonyl chloride, 2-propanesulfonyl chloride, isopropylsulphonyl chloride, isopropyl sulfonyl chloride, ccris 4139, 2-propanesulfonylchloride, isopropanesulfonyl chloride, iso-propylsulfonyl chloride, 2-propane sulfonyl chloride, isopropylsulphonyl choride
PubChem CID 82408
IUPAC Name propane-2-sulfonyl chloride
SMILES CC(C)S(=O)(=O)Cl

Specifications

Color Yellow
Boiling Point 83°C
Quantity 5 g
UN Number 3265
Formula Weight 142.60
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropylsulfonyl Chloride