accessibility menu, dialog, popup

UserName

H3CCH3SClOO

CAS RN 10147-37-2

IUPAC Name: propane-2-sulfonyl chloride
Synonyms: propane-2-sulfonyl chloride
Molecular Weight (g/mol): 142.6
Molecular Formula: C3H7ClO2S
InChi Key: DRINJBFRTLBHNF-UHFFFAOYSA-N
SMILES: CC(C)S(Cl)(=O)=O
Boiling Point 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
  • (2)
  • (5)
  • (1)
  • (3)
  • (3)
  • (2)