missing translation for 'onlineSavingsMsg'
Learn More

Isopropylhydrazine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7131683
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09641G 1 g
I09645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09641G Supplier TCI America Supplier No. I09641G

Chemical Identifiers

CAS 16726-41-3
Molecular Formula C3H11ClN2
Molecular Weight (g/mol) 110.585
MDL Number MFCD01746887
InChI Key ILULYDJFTJKQAP-UHFFFAOYSA-N
PubChem CID 85573
IUPAC Name propan-2-ylhydrazine;hydrochloride
SMILES CC(C)NN.Cl

Specifications

Melting Point 123°C
Color White
Quantity 1 g
Formula Weight 110.59
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Isopropylhydrazine Hydrochloride