accessibility menu, dialog, popup

UserName

HClCH3H3CNHH2N

CAS RN 16726-41-3

IUPAC Name: hydrogen (propan-2-yl)hydrazine chloride
Synonyms: hydrogen isopropylhydrazine chloride
Molecular Weight (g/mol): 110.59
Molecular Formula: C3H11ClN2
InChi Key: ILULYDJFTJKQAP-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC(C)NN
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results