missing translation for 'onlineSavingsMsg'
Learn More

Isopropyldimethylchlorosilane, 95%

Catalog No. AC313570250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC313570250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC313570250 Supplier Thermo Scientific Chemicals Supplier No. 313570250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3634-56-8 | C5H13ClSi | 136.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3634-56-8
Molecular Formula C5H13ClSi
Molecular Weight (g/mol) 136.69
MDL Number MFCD00009917
InChI Key YCXVDEMHEKQQCI-UHFFFAOYSA-N
Synonym isopropyldimethylchlorosilane, dimethylisopropylchlorosilane, chloro isopropyl dimethylsilane, chloro dimethyl isopropylsilane, chlorodimethylisopropylsilane, silane,chlorodimethyl 1-methylethyl, silane, chlorodimethyl 1-methylethyl, dmipscl, acmc-1bn7i, chlorodimethyl isopropylsilane
PubChem CID 5102883
IUPAC Name chloro-dimethyl-propan-2-ylsilane
SMILES CC(C)[Si](C)(C)Cl

Specifications

Color Colorless
Density 0.8700g/mL
Boiling Point 110.0°C to 114.0°C
Flash Point 15°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4150 to 1.4170 (20°C, 589nm)
Quantity 25 g
Specific Gravity 0.87
Formula Weight 136.7
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Isopropyldimethylchlorosilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

RUO – Research Use Only