missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropylcyclohexane 99.0+%, TCI America™
$96.94 - $691.57
Chemical Identifiers
| CAS | 696-29-7 |
|---|---|
| Molecular Formula | C9H18 |
| Molecular Weight (g/mol) | 126.24 |
| MDL Number | MFCD00001480 |
| InChI Key | GWESVXSMPKAFAS-UHFFFAOYSA-N |
| Synonym | Hexahydrocumene |
| PubChem CID | 12763 |
| IUPAC Name | (propan-2-yl)cyclohexane |
| SMILES | CC(C)C1CCCCC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
I016125ML
|
TCI America
I016125ML |
25 mL |
Each for $96.94
|
|
|||||
|
I0161500ML
|
TCI America
I0161500ML |
500 mL |
Each for $691.57
|
|
|||||
Chemical Identifiers
| 696-29-7 | |
| 126.24 | |
| GWESVXSMPKAFAS-UHFFFAOYSA-N | |
| 12763 | |
| CC(C)C1CCCCC1 |
| C9H18 | |
| MFCD00001480 | |
| Hexahydrocumene | |
| (propan-2-yl)cyclohexane |
Specifications
| 696-29-7 | |
| 151°C | |
| MFCD00001480 | |
| 3295 | |
| GWESVXSMPKAFAS-UHFFFAOYSA-N | |
| (propan-2-yl)cyclohexane | |
| 12763 | |
| ≥99.0% (GC) | |
| Isopropylcyclohexane |
| Colorless | |
| C9H18 | |
| 25 mL | |
| Hexahydrocumene | |
| CC(C)C1CCCCC1 | |
| 126.24 | |
| 126.24 | |
| Liquid |
Safety and Handling
TSCA : Yes