accessibility menu, dialog, popup

UserName

H3CCH3

CAS RN 696-29-7

IUPAC Name: (propan-2-yl)cyclohexane
Synonyms: isopropylcyclohexane
Molecular Weight (g/mol): 126.24
Molecular Formula: C9H18
InChi Key: GWESVXSMPKAFAS-UHFFFAOYSA-N
SMILES: CC(C)C1CCCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results